Please login to your online account to display your discounted pricing

Thermo Scientific™ ABsolute Blue qPCR Mixes


Make verification of master mix dispensing quick, easy and foolproof with these colored qPCR master mixes for probe chemistry using standard cycling protocols.

Manufacturer: thermo scientific™  AB4139/B

 View more versions of this product

 Disclaimers

Purchase of this product includes an immunity from suit under patents specified in the product insert to use only the amount purchased for the purchaser's own internal research. No other patent rights are conveyed expressly, by implication, or by estoppel. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.

Catalog No. AB4139B

  • / Pack of 10 Each
 

Description & Specifications

Specifications

Compatibility ABsolute Blue qPCR mixes are ideal for use with white reaction plates and tubes. Check qPCR cycler compatibility with our qPCR reagent selection guide.
For Use With (Application) Ideal for QPCR analysis in molecular biology applications, including gene expression and SNP typing
Includes ABsolute Blue qPCR Buffer (Thermo-Start DNA polymerase, dNTPs, MgCl2, ROX™ passive reference dye (only in ROX mixes); Separate ROX vial (only in kits indicated)
Product Type QPCR ROX Mix
Quantity 10 x 5mL
Storage Requirements Store at -20°C. Avoid freeze/thaw cycles. The ROX dye is light sensitive; exposure to light should be minimized for vials containing ROX dye.
Sufficient For 4000 x 25µL Rxns
Item Description ROX Mix; 4000; 10 x 400 x 25uL rxns
Product Name QPCR ROX Mix

ABsolute Blue qPCR Mixes are 2X master mixes for probe chemistry. There are suitable alternatives available for all common real-time instruments. ABsolute Blue qPCR Mixes utilize Thermo-Start Taq hot start enzyme for increased sensitivity. The master mixes incorporate an inert blue dye to enhance the contrast between reagent and plastic, making verification of master mix dispensing quick, easy, and foolproof.

Highlights

  • High performance - ABsolute sensitivity, consistency & convenience
  • Enhanced contrast and reduced errors - easily identify correctly aliquotted reaction mix in all plates and tubes
  • Universal - compatible with all major qPCR platforms
  • Adaptable - available with different ROX passive reference dye concentrations