Please login to your online account to display your discounted pricing

(±)-Abscisic acid, >98%, MP Biomedicals™


Quantity 500mg

Manufacturer: mp biomedicals  19067390

 View more versions of this product

Catalog No. ICN19067390

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Fine Powder
Formula Weight 264.3
Color White
Quantity 500mg
Chemical Name or Material Abscisic acid
Assay Percent Range ≥98%
Molecular Formula C15H20O4
Synonym (+)-cis, trans-Abscisic acid; 5-[1-Hydroxy-2,6,6-trimethyl-4-oxocyclohex-2-en-1-yl]-3-methyl-[2Z, 4E]-pentadienoic acid
Recommended Storage Store at -0°C.
CAS 14375-45-2
Storage Note 1 Protect from light, store under nitrogen.

Specifications

Chemical Name or Material Abscisic acid
CAS 14375-45-2
Physical Form Fine Powder
Assay Percent Range ≥98%
Molecular Formula C15H20O4
Formula Weight 264.3
Synonym (+)-cis, trans-Abscisic acid; 5-[1-Hydroxy-2,6,6-trimethyl-4-oxocyclohex-2-en-1-yl]-3-methyl-[2Z, 4E]-pentadienoic acid
Accessories

Store at -0°C.

Protect from light, store under nitrogen.