Please login to your online account to display your discounted pricing

(±)-Abscisic Acid 98%, ACROS Organics™


Quantity 500mg
Packaging Glass bottle

Manufacturer: fisher scientific acros  133485000

 View more versions of this product

Catalog No. AC133485000

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 264.32
Quantity 500mg
Merck Index 15, 12
TSCA TSCA
Sublimation Point 0120.0°C
CAS Max % 100.0
Synonym (2-cis, 4-trans)-5-(1-Hydroxy-2, 6
Chemical Name or Material (±)-Abscisic acid
Assay Percent Range 98%
Molecular Formula C15H20O4
CAS 14375-45-2
Packaging Glass bottle
CAS Min % 97.5
Name Note 98%
MDL Number MFCD00075619
Health Hazard 1 LIGHT SENSITIVE

Specifications

Chemical Name or Material (±)-Abscisic acid
Name Note 98%
CAS 14375-45-2
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C15H20O4
Formula Weight 264.32
MDL Number MFCD00075619
Synonym (2-cis, 4-trans)-5-(1-Hydroxy-2, 6
Accessories

LIGHT SENSITIVE

SDS & Safety Documents