Please login to your online account to display your discounted pricing

(±)-Abscisic Acid 98%, ACROS Organics™

$65.08 - $229.43

Chemical Identifiers

Chemical Name or Material (±)-Abscisic acid
Name Note 98%
CAS 14375-45-2
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C15H20O4
Formula Weight 264.32
MDL Number MFCD00075619
Synonym (2-cis, 4-trans)-5-(1-Hydroxy-2, 6
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC133485000

  SDS
fisher scientific acros
133485000
Glass bottle 500mg Each for $229.43

AC133481000

  SDS
thermo fisher scientific
AC133481000
Glass bottle 100mg Each for $65.08
Description & Specifications

Specifications

Formula Weight 264.32
Quantity 500mg
Merck Index 15, 12
TSCA TSCA
Sublimation Point 0120.0°C
CAS Max % 100.0
Synonym (2-cis, 4-trans)-5-(1-Hydroxy-2, 6
Packaging Glass bottle
CAS 14375-45-2
Chemical Name or Material (±)-Abscisic acid
Molecular Formula C15H20O4
Assay Percent Range 98%
CAS Min % 97.5
MDL Number MFCD00075619
Name Note 98%
Health Hazard 1 LIGHT SENSITIVE
Safety and Handling

LIGHT SENSITIVE