missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Abnova™ Serotonin ELISA Kit
Enzyme immunoassay for the quantitative determination of Serotonin in serum, urine and platelets
$960.15 - $960.15
Specifications
Detection Method | Colorimetric |
---|---|
Quantity | 1 Kit |
For Use With (Application) | Quantification |
Catalog Number | Mfr. No. | For Use With (Application) | Quantity | Sample Type | Sample Volume | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With (Application) | Quantity | Sample Type | Sample Volume | Price | Quantity | ||
89-100-615
|
Abnova™
KA2518 |
Quantification | 1 Kit | Biological sample | 100 μL |
Each for $960.15
|
|
||
Description
- Sample Type: Platelet, Serum, Urine
- Sample Volume: 25μL
Applications:
Quantification
Specifications
Colorimetric | |
1 Kit | |
Quantification |