missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Abnova ROS1 Split FISH Probe

$2,934.32 - $2,934.32

Specifications

Probe Split FISH Probe
Storage Requirements Store at 4°C in the dark.
View More Specs

Includes: DAPI Counterstain (1500ng/mL) 250μL


Products ${productFamilyLength}
Catalog Number Mfr. No. For Use With (Application) Price Quantity  
Catalog Number Mfr. No. For Use With (Application) Price Quantity  
89-028-836 Abnova Corporation
FS0015
Fluorescent in situ hybridization Each for $2,934.32
Specifications

Description

Labeled FISH probes for identification of gene split using Fluoresecent In Situ Hybridization Technique.

  • Probe1: ROS1(Texas Red)
  • Size: Approximately 250kb
  • Fluorophore: Texsas Red
  • Location: 6q22.1
  • Probe2: ROS1(FITC)
  • Size:Approximately 720kb
  • Fluorophore: FITC
  • Location: 6q22.1
  • Origin: Human
  • Source: Genomic DNA

Fluorescent In Situ Hybridization (Cell)

Description & Specifications

Specifications

Split FISH Probe
Store at 4°C in the dark.