missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Abnova™ MB (Human) ELISA Kit

$446.11 - $446.11

Specifications

Calibration Range 0 to 1000ng/mL
Detection Limit 5ng/mL
Detection Method Colorimetric
Conjugate HRP
Cross Reactivity Human
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Product Type Price Quantity  
Catalog Number Mfr. No. Product Type Price Quantity  
89-100-015 Abnova™
KA0237
ELISA Kit Each for $446.11
Specifications Description & Specifications

Specifications

0 to 1000ng/mL
Colorimetric
Human
Serum
5ng/mL
HRP
Antibody-Coated Wells 96 wells, Reference Standard Set Contains 0, 25, 100, 250, 500, and 1000 ng/ml in. 0.5mL each, Sample Diluent (25mL), Enzyme Conjugate Reagent (22mL), TMB Reagent (One-Step) (11mL), Stop Solution (1N HCl) (11mL)
20μL