missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Abnova™ Iron Assay Kit
To measure ferrous and/or ferric ion in samples, functional study
Supplier: Abnova™ KA0814
Includes:
- Iron Assay Buffer 15mL
- Iron Probe 12mL
- Iron Reducer 0.7mL
- Iron Standard (100 mM) 0.1mL
Specifications
8 to 400μM | |
Iron Assay Kit is used for measuring ferrous and/or ferric ion in samples. | |
Functional Study | |
1 Kit | |
Iron Assay Kit |
Store the kit at -20°C. | |
Colorimetric | |
15mL Iron assay buffer, 12mL Iron probe, 0.7mL Iron reducer, 0.1mL Iron standard (100mM) | |
Biological Fluid, Cell Lysate, Culture Medium, Serum, Tissue Lysate, Urine |