Please login to your online account to display your discounted pricing

Abnova™ IL6R (Human) ELISA Kit

$962.01 - $962.01

Specifications

Detection Method Colorimetric
Cross Reactivity Human
Storage Requirements Store the kit at 4°C.
Sufficient For 96 Assays
View More Specs
Products
Catalog Number Mfr. No. Calibration Range Includes Sample Volume Conjugate Price Quantity    

89-022-605

 
abnova™
KA0124
0.08 to 5ng/mL Antibody coated microwells (96 wells), Biotin-Conjugate antibody (100μL), Streptavidin-HRP (150μL), Standard (lyophilized) 2 vials, Sample Diluent (12mL), Assay Buffer Concentrate 20x (5mL), Wash Buffer Concentrate 20X (50mL), Substrate Solution (15mL), Stop Solution (15mL), Blue-Dye (0.4mL), Green-Dye (0.4mL), Red-Dye (0.4mL) 20μL (1:50 prediluted) HRP Each for $962.01

89-022-950

 
abnova™
KA0523
31.2 to 2000pg/mL Antibody coated microwells (96 wells), BIOTIN CONJUGATE (7mL), AVIDIN CONJUGATE (12mL), IL-6 sR STANDARD 4ng/vial x 2, CALIBRATOR DILUENT I (5X) (25mL), CALIBRATOR DILUENT II (25mL), WASH BUFFER (20X) (60mL), SUBSTRATE A (10mL), SUBSTRATE B (10mL), Stop Solution (14mL) 100μL HRP Each for $962.01
Description & Specifications

Specifications

Detection Method Colorimetric
Cross Reactivity Human
Storage Requirements Store the kit at 4°C.
Sufficient For 96 Assays