Please login to your online account to display your discounted pricing

Abnova™ IL6 (Human) ELISA Kit

$941.69 - $941.69

Specifications

Detection Method Colorimetric
View More Specs
Includes: 
  • Antibody coated microwells 96 wells
  • Biotin-Conjugate antibody 100μL
  • Streptavidin-HRP 150μL
  • Standard (lyophilized) 2 vials
  • Sample Diluent 12mL
  • Assay Buffer Concentrate 20x 5mL
  • Wash Buffer Concentrate 20x 50mL
  • Substrate Solution 15mL
  • Stop Solution 15mL
  • Blue-Dye 0.4mL
  • Green-Dye 0.4mL
  • Red-Dye 0.4mL
Products
Catalog Number Mfr. No. Calibration Range Sample Type Storage Requirements Product Type Price Quantity    

89-022-604

 
abnova corporation
KA0123
1.56 to 100pg/mL Cell Culture Supernatant, Plasma (Citrate, EDTA, Heparin), Serum 4°C IL-6 (Human) ELISA kit Each for $941.69
Description & Specifications

Specifications

Detection Method Colorimetric

IL6 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human IL-6.

Quantification