Please login to your online account to display your discounted pricing

Abnova™ Helicobacter pylori IgG ELISA Kit

$436.68 - $436.68

Specifications

Detection Limit 99.00%
Detection Method Colorimetric
Conjugate HRP
Cross Reactivity Viruses
Product Type ELISA Kit
View More Specs
Products
Catalog Number Mfr. No. Includes Price Quantity    

89-100-029

 
abnova™
KA0220
Purified H. pylori antigen coated microtiter plate (96 wells), Enzyme Conjugate Reagent (red color): 13mL, Sample Diluent (green color): 22mL, H. pylori Negative Control, <6.25U/ml 100 (100μL), H. pylori Standards, 0, 6.25, 12.5, 25, 50, and 100U/mL (100μL each), H. pylori Positive Control, >100U/mL (100μL), Wash Buffer (20): 50mL, TMB Reagent (One-Step): 11mL, Stop Solution (1N HCl): 11mL Each for $436.68
Description & Specifications

Specifications

Detection Limit 99.00%
Detection Method Colorimetric
Conjugate HRP
Cross Reactivity Viruses
Product Type ELISA Kit
Regulatory Status RUO
Sample Type Serum
Storage Requirements 4°C
Sufficient For 96 Assays
Volume (Metric) Sample 100μL