missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Abnova™ Glucose Assay Kit
To measure glucose, functional study
Supplier: Abnova™ KA0831
Includes:
- Glucose assay buffer, 25mL
- Glucose probe (in DMSO), 0.2mL
- Glucose enzyme mix (lyophilized), 1 vial
- Glucose standard (100 nmol/μL), 10μL
Specifications
1 to 10000μM | |
Glucose Assay Kit is used for measuring glucose. | |
Functional Study | |
1 Kit | |
Glucose Assay Kit |
Store the kit at -20°C. | |
Colorimetric, Fluorometric | |
25mL Glucose assay buffer, 0.2mL Glucose probe (in DMSO), 1 vial Glucose enzyme mix (lyophilized), 100μL Glucose standard (100nM/μL) | |
Biological Fluid, Cell Culture Supernatant, Growth Medium, Plasma, Serum, Tissue Culture Supernatant, Urine |