missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Abnova™ COMP (Human) ELISA Kit

$973.18 - $973.18

Specifications

Detection Method Colorimetric
Cross Reactivity Human
Product Type ELISA Kit
Regulatory Status RUO
Sufficient For 96 Assays
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Calibration Range Detection Limit Storage Requirements Conjugate Price Quantity  
Catalog Number Mfr. No. Calibration Range Detection Limit Storage Requirements Conjugate Price Quantity  
89-100-009 Abnova™
KA0021
4 to 128ng/mL 0.4ng/mL 4°C HRP Each for $973.18
Specifications Description & Specifications

Specifications

Colorimetric
ELISA Kit
96 Assays
Human
RUO