missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Abnova™ Carbohydrate Assay Kit (Colorimetric)
Quantitative colorimetric determination of carbohydrate at 490nm.
Supplier: Abnova™ KA3756
Specifications
Store the kit at 4°C. | |
Colorimetric | |
1 Kit |
Carbohydrate Assay Kit is a quantitative colormetric determination of carbohydrate at 490 nm. | |
Functional Study | |
Attached Cell, Beverage, Food Product, Plant Tissue, Suspension Cell, Tissue |