missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Abnova™ Calcium Assay Kit (Colorimetric)
To provide a simple assay in the physiologically important range of calcium concentration, functional study
Supplier: Abnova™ KA0812
Includes:
- Calcium assay buffer, 15mL
- Chromogenic reagent, 25mL
- Calcium standard (500mm), 100μL
Specifications
0.1 to 25mm | |
Calcium Assay Kit (Colorimetric) is used for providing a simple assay in the physiologically important range of calcium concentration. | |
Functional Study | |
1 Kit | |
Calcium Assay Kit |
Store the kit at 4°C. | |
Colorimetric | |
15mL calcium assay buffer, 25mL chromogenic reagent, 100μL calcium standard (500mM) | |
Biological Fluid, Cell Lysate, Culture Medium, Serum, Tissue Lysate, Urine |