missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Abnova™ Alcohol Dehydrogenase Assay Kit (Colorimetric)
Quantitative colorimetric determination of alcohol dehydrogenase at 565nm.
Supplier: Abnova™ KA3785
Specifications
Store the kit at -20°C. | |
Colorimetric | |
1 Kit | |
20 μL |
Alcohol dehydrogenase Assay Kit is a quantitative colorimetric determination of alcohol dehydrogenase at 565 nm. | |
Functional Study | |
Culture Medium, Plasma, Serum, Tissue, Urine |