missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Abnova™ ACTN4 DNA Probe

Labeled ACTN4 DNA Probe for Hybridization

Manufacturer:  Abnova™ DF0001

Catalog No. 02-990-406



Description

Description

  • Applications include DNA hybridization
  • Human genomic DNA
  • Analytical and performance characteristics are not established
  • Store at 3-5°C in the dark; please aliquot before use

Specifications

Specifications

Store at 3-5°C in the dark. Please aliquot before use.
ACTN4 DNA Probe
Labeled ACTN4 DNA Probe for Hybridization
200 μL
DNA hybridization
SDS
Documents

Documents

Product Certifications
Promotions

Promotions