ABL2 (Arg) Recombinant Human Protein DFS Item
FLT1 (VEGFR1) Recombinant Human Protein
Manufacturer:  Life Technologies LSPV3266
 View more versions of this product
Catalog No. PV3266
Description
Description
ABL2 (Arg) is an Abl-related non-receptor protein tyrosine kinase that links cell surface receptors to cell proliferation and⁄or cytoskeletal reorganization.
Specifications
Specifications
NP_009298.1
Pharma and biopharma, protein assays and analysis, target and lead identification and validation
130.9kDa
70% by SDS-PAGE
–80°C
1 Each
ABL2 (Arg)
ABL2 (ARG) was pre-diluted in enzyme dilution buffer and assayed in 50mM Tris (pH 7.5), 10mM MgCl2, 1mM DTT, 0.01% NP-40, 200μM ATP, 100μM Abl1 peptide substrate (EAIYAAPFAKKK) and trace [32P]-γ-ATP for 10 minutes at 30°C
Kinase and inhibitors
ABL
2,250nmoles of phosphate transferred to Abl1 peptide substrate (EAIYAAPFAKKK) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 1μg/mL
His Tag
0.35mg/mL
27
ABL2 (Arg) Recombinant Human Protein
10μg
For research use only. Not for use in diagnostic procedures
Cytoplasmic tyrosine kinase
ABL2 (Arg) Recombinant Human Protein
Kinase assay
Baculovirus
ABL2 (Arg) Recombinant Human Protein
Human
Documents
For Research Use Only. Not for use in diagnostic procedures.