missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
ABL1 [T315I] Recombinant Human Protein, Invitrogen™
The T315I mutant is a Gleevec resistant form of ABL1.
$2650.00 - $11260.00
Specifications
Accession Number | NP_005148.2, T315I |
---|---|
For Use With (Application) | Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology |
Formulation | 50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% NP-40, 2mM DTT and 50% glycerol |
Molecular Weight (g/mol) | 125.4kDa |
Storage Requirements | Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
PR7429B
|
Thermo Fisher Scientific
PR7429B |
100μg |
Each for $2,650.00
|
|
|||||
PV4184
|
Thermo Fisher Scientific
PV4184 |
1mg |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
The ABL1 proto-oncogene encodes a cytoplasmic and nuclear protein tyrosine kinase that has been implicated in processes of cell differentiation, cell division, cell adhesion, and stress response.
Specifications
NP_005148.2, T315I | |
50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% NP-40, 2mM DTT and 50% glycerol | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
ABL Kinase Family | |
Recombinant | |
Baculovirus |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
125.4kDa | |
RUO - research use only | |
ABL1 T315I, ABL, c-ABL | |
Human | |
His Tag | |
Frozen |