ABL1 [T315I] Recombinant Human Protein, Invitrogen™ DFS Item
The T315I mutant is a Gleevec resistant form of ABL1.
Manufacturer:  Life Technologies LSPV4184
 View more versions of this product
Catalog No. PV4184
Description
Description
The ABL1 proto-oncogene encodes a cytoplasmic and nuclear protein tyrosine kinase that has been implicated in processes of cell differentiation, cell division, cell adhesion, and stress response.
Specifications
Specifications
NP_005148.2, T315I
Frozen
125.4kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
Kinase Assay
ABL Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% NP-40, 2mM DTT and 50% glycerol
1mg
RUO - research use only
ABL1 T315I, ABL, c-ABL
Baculovirus
Human
His Tag
Documents