ABL1 Recombinant Human Protein DFS Item
ABL1 Recombinant Human Protein
Manufacturer:  Life Technologies LSPV3585
Catalog No. PV3585
Description
Description
The ABL1 proto-oncogene encodes a cytoplasmic and nuclear protein tyrosine kinase that has been implicated in processes of cell differentiation, cell division, cell adhesion, and stress response.
Specifications
Specifications
NP_005148.2
Protein assays and analysis
Frozen
ABL1 Recombinant Human Protein
1mg
For research use only. Not for use in diagnostic procedures
ABL1, ABL, c-ABL
ABL1 was pre-diluted in enzyme dilution buffer and assayed in 50mM Tris (pH 7.5), 10mM MgCl2, 1mM DTT, 0.01% NP-40, 200μM ATP, 100μM Abl1 peptide substrate (EAIYAAPFAKKK) and trace [32P]-γ-ATP for 10 minutes at 30°C
Baculovirus
ABL1 Recombinant Human Protein
Human
0.2mg/mL
Frozen
125.4kDa
80% by SDS-PAGE
–80°C
1 Each
ABL1 Recombinant Human Protein
Kinase assay
ABL
2090nmoles of phosphate transferred to Abl1 peptide substrate (EAIYAAPFAKKK) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 0.4μg/mL
His Tag
Documents
For Research Use Only. Not for use in diagnostic procedures.