ABL1 [M351T] Recombinant Human Protein, Invitrogen™ DFS Item
Human ABL1, internal fragment, amino acids P118-S535 (as in GenBank entry NM_005157.3), M351T mutant, N-terminal GST-HIS6 fusion protein with a 3C cleavage site, expressed in Sf9 insect cells.
$614.00 - $9,380.00
Specifications
Host Species Human
Protein Tag GST Tag, His Tag
Recombinant Yes
View More Specs
Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PV6151 DFS Item
View Documents
Life Technologies
LSPV6151
10μg
Each for $614.00
PV6152 DFS Item
View Documents
Life Technologies
LSPV6152
100μg
Each for $2,095.00
PV6153 DFS Item
View Documents
Life Technologies
LSPV6153
1mg
Each for $9,380.00
Description
Description
  • Validated in LanthaScreen™ Kinase Binding Assay
  • Protocols available for simple use and optimized performance
  • Multiple pack sizes available for convenient use
Specifications
Specifications
Human
GST Tag, His Tag
Yes
SDS