Learn More
Abietic acid, 90+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 042582MD
Description
Abietic acid is used to make ester gums in lacquers, varnishes and soaps. It is also used in metal resonates and plastics. Further, it is utilized in musical instruments to make them less slippery. It acts as a precursor to natural product synthesis. In addition to this, it is used to depackage integrated circuits from their epoxy coatings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Abietic acid | |
514-10-3 | |
C20H30O2 | |
2221451 | |
abietic acid, sylvic acid, abietate, l-abietic acid, rosin acid, 7,13-abietadien-18-oic acid, kyselina abietova, kyselina abietova czech, unii-v3dhx33184, ccris 3183 | |
RSWGJHLUYNHPMX-ONCXSQPRSA-N | |
(1R,4aR,4bR,10aR)-1,4a-dimethyl-7-propan-2-yl-2,3,4,4b,5,6,10,10a-octahydrophenanthrene-1-carboxylic acid | |
10569 | |
302.45 | |
250 mg | |
Crystalline |
148°C to 152°C | |
Yellow | |
MFCD03423567 | |
14,7 | |
Soluble in acetone,petroleum ether,diethyl ether and ethanol. Insoluble in water. | |
CC(C)C1=CC2=CC[C@@H]3[C@](C)(CCC[C@@]3(C)C(O)=O)[C@H]2CC1 | |
302.46 | |
CHEBI:28987 | |
≥90% | |
Characteristic | |
Air sensitive |
Chemical Identifiers
514-10-3 | |
302.46 | |
RSWGJHLUYNHPMX-ONCXSQPRSA-N | |
10569 | |
(1R,4aR,4bR,10aR)-1,4a-dimethyl-7-propan-2-yl-2,3,4,4b,5,6,10,10a-octahydrophenanthrene-1-carboxylic acid |
C20H30O2 | |
MFCD03423567 | |
abietic acid, sylvic acid, abietate, l-abietic acid, rosin acid, 7,13-abietadien-18-oic acid, kyselina abietova, kyselina abietova czech, unii-v3dhx33184, ccris 3183 | |
CHEBI:28987 | |
CC(C)C1=CC2=CC[C@@H]3[C@](C)(CCC[C@@]3(C)C(O)=O)[C@H]2CC1 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
DOTInformation : Hazard Class: 9; Packaging Group: III
EINECSNumber : 208-178-3
RTECSNumber : TP8580000
TSCA : Yes
storageNote1 : 2°C to 8°C
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only