missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

384-Well Full Skirted Plate, Extra Volume

Perform post-PCR sequencing and wash steps in the larger wells of this 384-well PCR plate with a 40 µL well volume.

Manufacturer:  Abgene Inc Usa AB0937W

 View more versions of this product

Catalog No. AB-0937W


 

Specifications

Description

Extra Volume 384-well full-skirted plate permits enables post-PCR applications such as enzymatic digestions and ethanol precipitation to be carried out in the wells due to the increased well volume.

  • Tall chimney well design gives working well volume of 40μL, a 60% volume increase over standard 384-well PCR plates
  • Increased well volume allows for post-PCR applications such as enzymatic digestions and ethanol precipitation to be carried out directly in the same plate
  • Raised rim design around each well enables secure sealing and safeguards against evaporation
  • Uniform, thin well walls deliver maximum and consistent heat transfer for equally high performance from every sample
  • SBS footprint
  • 55μL maximum well volume, 40μL working volume
  • Cut Corner: A24

Industry-leading Manufacturing and QC Testing for ultimate product reliability:

  • Cleanroom Produced - To avoid particulate and biological contaminants that can interfere with molecular biology applications
  • Virgin Medical Grade Polypropylene - Specialized polymer is highly inert and will not interfere with PCR reaction components
  • Integrity Testing - Every well of every plate is visually inspected and tested using an electrostatic pinhole detection method
  • PCR Functional Assays - Products are tested in PCR assays to verify clean negative controls and consistent technical replicates
  • Evaporation Testing - Samples from each lot are run through PCR cycling to test sealing performance
  • Biological Testing - All lots are biologically tested to certify them free of RNase, DNase, and DNA

Format Options:

Bar Coded Plates

  • Feature two industry standard 128 bar codes carefully designed and positioned for compatibility with all major bar code readers
  • Codes are random and each bar code also includes a human readable format
  • Wide temperature tolerance (-196° to +120°C)
  • Proprietary coating for superior scratch-resistance
  • Precise sizing and placement for reliable scanning
  • Custom bar coding also available, please enquire

White Plates

  • Designed specifically for qPCR
  • Reflect significantly more signal than traditional clear plates, which ensures that even the lowest levels of fluorescence are detected
  • Enable consistent signal reflection - the opaque well walls prevent signal from passing through to the cycler block where it can be inconsistently reflected or absorbed, keeping variations in the cycler block from affecting your qPCR data

Colored Plates

  • Great for sample organization and tracking
  • Available in five distinct colors: Blue, Green, Purple, Red, and Yellow

Compatible with:

Adhesive Sealing

Recommended for:

PCR; qPCR / Real-Time PCR

Description & Specifications

Specifications

Certified free of human DNA, RNase, DNase, and endotoxins.
Full Skirted
55µL
White
384
40µL