Please login to your online account to display your discounted pricing
Thermo Scientific™ ABgene™ 24-Well PCR Plates, Semi-Skirted
Optimize PCR and qPCR applications with these 24-well semi-skirted plates.
Manufacturer: thermo scientific™  AB0624B
 View more versions of this product
Catalog No. AB-0624B
  • / Pack of 50 Each
Description & Specifications
Specifications
Description Semi-Skirted
No. of Wells 24-Well (3 x 8 wells)
Volume (Metric) Well 300µL
Color Blue
24-Well Semi-Skirted PCR Plate is pre-cut into 24-well segments. Its convenient format allows for lower sample numbers. The semi skirt adds rigidity and allots space for labeling or barcoding. Available in various colors and compatible with standard multichannel pipettes. Sealing is possible with adhesive films and foils and all cap strips.
  • Conveniently pre-cut into 24-well segments
  • Semi-skirt adds rigidity and allows for labeling or bar coding
  • Thin wall polypropylene