Please login to your online account to display your discounted pricing

AA 2414, Tocris

Thromboxane A2 (TP) receptor antagonist

$181.19 - $762.58

Chemical Identifiers

Chemical Name or Material AA 2414
Assay Percent Range >98%
Molecular Formula C22H26O4
Formula Weight 354.44
Synonym 7-(3,5,6-Trimethyl-1,4-benzoquinon-2-yl)-7-phenylheptanoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

302510

 
r & d systems
RDS302510
10mg Each for $181.19

302550

 
r & d systems
RDS302550
50mg Each for $762.58
Description & Specifications

Specifications

Formula Weight 354.44
Quantity 10mg
Chemical Name or Material AA 2414
Assay Percent Range >98%
Molecular Formula C22H26O4
CAS 112665-43-7
Synonym 7-(3,5,6-Trimethyl-1,4-benzoquinon-2-yl)-7-phenylheptanoic acid
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C