missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

A20, Acidic Cytokeratin Mouse, Clone: AE1, eBioscience™

Mouse Monoclonal Antibody

$170.28 - $394.68

Specifications

Antigen A20, Acidic Cytokeratin
Clone AE1
Host Species Murine
Gene Alias keratin
Applications Immunocytochemistry,Immunohistochemistry,Immunohistochemistry (Formalin/Paraffin),Immunohistochemistry (Frozen),Western Blot
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5013762 Life Technologies
LS14900180
25μg Each for $170.28
5013763 Life Technologies
LS14900182
100μg Each for $394.68
Specifications

Description

The monoclonal antibody AE1 recognizes many members of the acidic (Type I) family of cytokeratins. Cytokeratins are intermediate filament proteins comprising one component of the cytoskeleton. There are two large families of cytokeratins, acidic and basic, but all contain the same basic domains (i.e. an α-helical core with an N- and C-terminal domain). The proteins are expressed in epithelial cells, but are developmentally regulated. Many tumors also express these cytokeratins, and their expression can help identify the origin of a neoplasm.

The AE1 monoclonal antibody recognizes 56.5, 54', 50, 50’, 48, and 40kDa proteins (also known as CK10, 14, 15, 16 and 19) in mouse, human, rat, primate (cynomolgus and rhesus), dog, cat, rabbit and chicken.
Description & Specifications

Specifications

A20, Acidic Cytokeratin
Murine
Immunocytochemistry,Immunohistochemistry,Immunohistochemistry (Formalin/Paraffin),Immunohistochemistry (Frozen),Western Blot
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL
AE1
keratin
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer