Please login to your online account to display your discounted pricing

A 967079, Tocris™

Selective TRPA1 channel blocker

$177.37 - $746.48

Chemical Identifiers

Chemical Name or Material A 967079
Assay Percent Range >98%
Molecular Formula C12H14FNO
Formula Weight 207.24
Synonym (1E, 3E)-1-(4-Fluorophenyl)-2-methyl-1-pentene-3-one oxime
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

471610

 
r & d systems
RDS471610
10mg Each for $177.37

471650

 
r & d systems
RDS471650
50mg Each for $746.48
Description & Specifications

Specifications

Formula Weight 207.24
Quantity 10mg
Molecular Formula C12H14FNO
Synonym (1E, 3E)-1-(4-Fluorophenyl)-2-methyl-1-pentene-3-one oxime
Assay Percent Range >98%
Recommended Storage Store at +4°C
CAS 1170613-55-4
Chemical Name or Material A 967079
Safety and Handling

Store at +4°C