Please login to your online account to display your discounted pricing

A 943931 dihydrochloride, Tocris™

Potent and selective H4 antagonist

$259.29 - $1,075.94

Chemical Identifiers

Chemical Name or Material A 943931 dihydrochloride
Assay Percent Range >98%
Molecular Formula C17H21N5·2HCl
Formula Weight 368.31
Synonym 4-((3R)-3-amino-pyrrolidin-1-yl)-6,7-dihydro-5H-benzo[6,7]cyclohepta[1,2-d]pyrimidin-2-ylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

375310

 
r & d systems
375310
10mg Each for $259.29

375350

 
r & d systems
375350
50mg Each for $1,075.94
Description & Specifications

Specifications

Formula Weight 368.31
Quantity 10mg
Synonym 4-((3R)-3-amino-pyrrolidin-1-yl)-6,7-dihydro-5H-benzo[6,7]cyclohepta[1,2-d]pyrimidin-2-ylamine
Assay Percent Range >98%
CAS 1227675-50-4
Chemical Name or Material A 943931 dihydrochloride
Molecular Formula C17H21N5·2HCl
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C