Please login to your online account to display your discounted pricing

A 839977, Tocris™

Potent P2X7 antagonist

$239.63 - $976.15

Chemical Identifiers

Chemical Name or Material A 839977
Assay Percent Range >98%
Molecular Formula C19H14Cl2N6O
Formula Weight 413.26
Synonym 1-(2, 3-Dichlorophenyl)-N-[[2-(2-pyridinyloxy)phenyl]methyl]-1H-tetrazol-5-amine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

423250

 
r & d systems
423250
50mg Each for $976.15

423210

 
r & d systems
423210
10mg Each for $239.63
Description & Specifications

Specifications

Formula Weight 413.26
Quantity 50mg
Chemical Name or Material A 839977
Assay Percent Range >98%
Molecular Formula C19H14Cl2N6O
Synonym 1-(2, 3-Dichlorophenyl)-N-[[2-(2-pyridinyloxy)phenyl]methyl]-1H-tetrazol-5-amine
CAS 870061-27-1
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C