Please login to your online account to display your discounted pricing

A 804598, Tocris™

Potent and selective P2X7 antagonist

$214.12 - $891.94

Chemical Identifiers

Chemical Name or Material A 804598
Assay Percent Range >99%
Molecular Formula C19H17N5
Formula Weight 315.37
Synonym N-Cyano-N''-[(1S)-1-phenylethyl]-N'-5-quinolinyl-guanidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

447310

 
r & d systems
RDS447310
10mg Each for $214.12

447350

 
r & d systems
RDS447350
50mg Each for $891.94
Description & Specifications

Specifications

Formula Weight 315.37
Quantity 10mg
Chemical Name or Material A 804598
Assay Percent Range >99%
Molecular Formula C19H17N5
CAS 1125758-85-1
Synonym N-Cyano-N''-[(1S)-1-phenylethyl]-N'-5-quinolinyl-guanidine
Recommended Storage Store at -20°C
Safety and Handling

Store at -20°C