Please login to your online account to display your discounted pricing

A 80426 mesylate, Tocris

High affinity α2 antagonist; Also 5-HT uptake inhibitor

$218.74 - $915.10

Chemical Identifiers

Chemical Name or Material A 80426 mesylate
Assay Percent Range >98%
Molecular Formula C23H27NO2·CH3SO3H
Formula Weight 445.57
Synonym N-Methyl-N-[[(1R)-1,2,3,4-tetrahydro-5-methoxy-1-naphthalenyl]methyl]-6-benzofuranethanamine mesylate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

234110

 
r & d systems
RDS234110
10mg Each for $218.74

234150

 
r & d systems
RDS234150
50mg Each for $915.10
Description & Specifications

Specifications

Formula Weight 445.57
Quantity 10mg
Chemical Name or Material A 80426 mesylate
Molecular Formula C23H27NO2·CH3SO3H
CAS 152148-64-6
Synonym N-Methyl-N-[[(1R)-1,2,3,4-tetrahydro-5-methoxy-1-naphthalenyl]methyl]-6-benzofuranethanamine mesylate
Recommended Storage Desiccate at +4°C
Assay Percent Range >98%
Safety and Handling

Desiccate at +4°C