missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

A 784168, Tocris™

Potent and selective TRPV1 antagonist

$244.81 - $997.23

Chemical Identifiers

Assay Percent Range >99%
Molecular Formula C19H15F6N3O3S
Formula Weight 479.4
Synonym 3, 6-Dihydro-3'-(trifluoromethyl)-N-[4-[(trifluoromethyl)sulfonyl]phenyl]-[1(2H), 2'-bipyridine]-4-carboxamide
Chemical Name or Material A 784168
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
431910 R & D Systems
RDS431910
10mg Each for $244.81
431950 R & D Systems
RDS431950
50mg Each for $997.23
Specifications Description & Specifications

Chemical Identifiers

>99%
479.4
A 784168
C19H15F6N3O3S
3, 6-Dihydro-3'-(trifluoromethyl)-N-[4-[(trifluoromethyl)sulfonyl]phenyl]-[1(2H), 2'-bipyridine]-4-carboxamide

Specifications

10mg
>99%
479.4
A 784168
824982-41-4
C19H15F6N3O3S
3, 6-Dihydro-3'-(trifluoromethyl)-N-[4-[(trifluoromethyl)sulfonyl]phenyl]-[1(2H), 2'-bipyridine]-4-carboxamide

Safety and Handling

Recommended Storage : Store at Room Temperature