Please login to your online account to display your discounted pricing

Tocris™ A 740003

Potent and selective P2X7 antagonist

$214.12 - $895.76

Specifications

Product Type A 740003
Inhibitors A 740003
Molecular Weight 474.55
Purity >98
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

370110R

 
tocris™
RDS370110
10mg Each for $214.12

370150R

 
tocris™
RDS370150
50mg Each for $895.76
Description & Specifications

Specifications

Product Type A 740003
Inhibitors A 740003
Molecular Weight 474.55
Purity >98

Potent and selective P2X7 receptor antagonist (IC50 values are 18 and 40nM for rat and human receptors respectively). Displays selectivity over a variety of P2X and P2Y receptors up to a concentration of 100μM. Reduces nociception in animal models of persistent neuropathic and inflammatory pain. Also reduces neuroblastoma tumor growth in mice.

  • Chemical Name: N-[1-[[(Cyanoamino)(5-quinolinylamino)methylene]amino]-2,2-dimethylpropyl]-3,4-dimethoxybenzeneacetamide
  • Formula: C26H30N6O3
  • CAS Number: 861393-28-4
  • Solubility: Soluble to 20mM in DMSO
  • Purity: >98