Please login to your online account to display your discounted pricing

A 61603 hydrobromide, Tocris

α1A agonist

$214.12 - $895.76

Chemical Identifiers

Chemical Name or Material A 61603 hydrobromide
Assay Percent Range >98%
Molecular Formula C14H19N3O3S·HBr
Formula Weight 390.29
Synonym N-[5-(4,5-Dihydro-1H-imidazol-2-yl)-2-hydroxy-5,6,7,8-tetrahydronaphthalen-1-yl]methanesulfonamide hydrobromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

105250

 
r & d systems
RDS105250
50mg Each for $895.76

105210

 
r & d systems
RDS105210
10mg Each for $214.12
Description & Specifications

Specifications

Chemical Name or Material A 61603 hydrobromide
Quantity 50mg
CAS 107756-30-9
Recommended Storage Desiccate at +4°C
Assay Percent Range >98%
Molecular Formula C14H19N3O3S·HBr
Formula Weight 390.29
Synonym N-[5-(4,5-Dihydro-1H-imidazol-2-yl)-2-hydroxy-5,6,7,8-tetrahydronaphthalen-1-yl]methanesulfonamide hydrobromide
Safety and Handling

Desiccate at +4°C