Please login to your online account to display your discounted pricing

A 61603 hydrobromide, Tocris


α1A agonist

Quantity 10mg

Manufacturer: r & d systems  RDS105210

 View more versions of this product

Catalog No. 105210

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 390.29
Quantity 10mg
CAS 107756-30-9
Assay Percent Range >98%
Recommended Storage Desiccate at +4°C
Molecular Formula C14H19N3O3S·HBr
Chemical Name or Material A 61603 hydrobromide
Synonym N-[5-(4,5-Dihydro-1H-imidazol-2-yl)-2-hydroxy-5,6,7,8-tetrahydronaphthalen-1-yl]methanesulfonamide hydrobromide

Specifications

Chemical Name or Material A 61603 hydrobromide
Assay Percent Range >98%
Molecular Formula C14H19N3O3S·HBr
Formula Weight 390.29
Synonym N-[5-(4,5-Dihydro-1H-imidazol-2-yl)-2-hydroxy-5,6,7,8-tetrahydronaphthalen-1-yl]methanesulfonamide hydrobromide
Accessories

Desiccate at +4°C