Please login to your online account to display your discounted pricing

A 582941, Tocris™

Partial agonist at α7 nAChR

$252.39 - $1,050.42

Chemical Identifiers

Chemical Name or Material A 582941
Assay Percent Range >98%
Molecular Formula C17H20N4
Formula Weight 280.37
Synonym Octahydro-2-methyl-5-(6-phenyl-3-pyridazinyl)-pyrrolo[3, 4-c]pyrrole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

434150

 
r & d systems
434150
50mg Each for $1,050.42

434110

 
r & d systems
434110
10mg Each for $252.39
Description & Specifications

Specifications

Formula Weight 280.37
Quantity 50mg
Chemical Name or Material A 582941
Assay Percent Range >98%
Molecular Formula C17H20N4
CAS 848591-90-2
Synonym Octahydro-2-methyl-5-(6-phenyl-3-pyridazinyl)-pyrrolo[3, 4-c]pyrrole
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature