Please login to your online account to display your discounted pricing

A 484954, Tocris™

CaM kinase III (eEF-2 kinase) inhibitor

$244.81 - $997.23

Chemical Identifiers

Chemical Name or Material A 484954
Assay Percent Range >98%
Molecular Formula C13H15N5O3
Formula Weight 289.29
Synonym 7-Amino-1-cyclopropyl-3-ethyl-1,2,3,4-tetrahydro-2,4-dioxopyrido[2,3-d]pyrimidine-6-carboxamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

448350

 
r & d systems
448350
50mg Each for $997.23

448310

 
r & d systems
448310
10mg Each for $244.81
Description & Specifications

Specifications

Formula Weight 289.29
Quantity 50mg
Chemical Name or Material A 484954
Assay Percent Range >98%
Molecular Formula C13H15N5O3
CAS 142557-61-7
Synonym 7-Amino-1-cyclopropyl-3-ethyl-1,2,3,4-tetrahydro-2,4-dioxopyrido[2,3-d]pyrimidine-6-carboxamide
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C