Please login to your online account to display your discounted pricing

A 438079 hydrochloride, Tocris

Competitive P2X7 antagonist

$214.12 - $895.76

Chemical Identifiers

Chemical Name or Material A 438079 hydrochloride
Assay Percent Range >98%
Molecular Formula C13H9Cl2N5·HCl
Formula Weight 342.61
Synonym 3-[[5-(2, 3-Dichlorophenyl)-1H-tetrazol-1-yl]methyl]pyridine hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

297210

 
r & d systems
RDS297210
10mg Each for $214.12

297250

 
r & d systems
RDS297250
50mg Each for $895.76
Description & Specifications

Specifications

Formula Weight 342.61
Quantity 10mg
Assay Percent Range >98%
CAS 899431-18-6
Recommended Storage Desiccate at Room Temperature
Chemical Name or Material A 438079 hydrochloride
Molecular Formula C13H9Cl2N5·HCl
Synonym 3-[[5-(2, 3-Dichlorophenyl)-1H-tetrazol-1-yl]methyl]pyridine hydrochloride
Safety and Handling

Desiccate at Room Temperature