Please login to your online account to display your discounted pricing

A 412997 dihydrochloride, Tocris™

Selective D4 agonist

$175.83 - $714.82

Chemical Identifiers

Chemical Name or Material A 412997 dihydrochloride
Assay Percent Range >98%
Molecular Formula C19H23N3O·2HCl
Formula Weight 382.33
Synonym N-(3-Methylphenyl)-4-(2-pyridinyl)-1-piperidineacetamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

455250

 
r & d systems
RDS455250
50mg Each for $714.82

455210

 
r & d systems
RDS455210
10mg Each for $175.83
Description & Specifications

Specifications

Formula Weight 382.33
Quantity 50mg
Chemical Name or Material A 412997 dihydrochloride
Assay Percent Range >98%
CAS 1347744-96-0
Synonym N-(3-Methylphenyl)-4-(2-pyridinyl)-1-piperidineacetamide
Molecular Formula C19H23N3O·2HCl
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature