Please login to your online account to display your discounted pricing

A 350619 hydrochloride, Tocris

Soluble guanylyl cyclase (sGC) activator

$225.77 - $924.23

Chemical Identifiers

Chemical Name or Material A 350619 hydrochloride
Assay Percent Range >99%
Molecular Formula C21H25ClN2OS·HCl
Formula Weight 425.41
Synonym 3-[2-[(4-Chlorophenyl)thiophenyl]-N-[4-(dimethylamino)butyl]-2-propenamide hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

275310

 
r & d systems
RDS275310
10mg Each for $225.77

275350

 
r & d systems
RDS275350
50mg Each for $924.23
Description & Specifications

Specifications

Formula Weight 425.41
Quantity 10mg
Chemical Name or Material A 350619 hydrochloride
Molecular Formula C21H25ClN2OS·HCl
Assay Percent Range >99%
CAS 1217201-17-6
Synonym 3-[2-[(4-Chlorophenyl)thiophenyl]-N-[4-(dimethylamino)butyl]-2-propenamide hydrochloride
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C