Please login to your online account to display your discounted pricing

A 286982, Tocris™

Potent inhibitor of the LFA-1/ICAM-1 interaction

$257.85 - $1,073.10

Chemical Identifiers

Chemical Name or Material A 286982
Assay Percent Range >99%
Molecular Formula C24H27N3O4S
Formula Weight 453.55
Synonym (2E)-1-(4-Acetyl-1-piperazinyl)-3-[4-[[2-(1-methylethyl)phenyl]thio]-3-nitrophenyl]-2-propen-1-one
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

422850

 
r & d systems
422850
50mg Each for $1,073.10

422810

 
r & d systems
422810
10mg Each for $257.85
Description & Specifications

Specifications

Formula Weight 453.55
Quantity 50mg
Chemical Name or Material A 286982
Assay Percent Range >99%
Molecular Formula C24H27N3O4S
Synonym (2E)-1-(4-Acetyl-1-piperazinyl)-3-[4-[[2-(1-methylethyl)phenyl]thio]-3-nitrophenyl]-2-propen-1-one
Recommended Storage Store at Room Temperature
CAS 280749-17-9
Safety and Handling

Store at Room Temperature