Please login to your online account to display your discounted pricing

A 1120, Tocris™

High affinity retinol-binding protein 4 (RBP4) ligand

$214.12 - $891.94

Chemical Identifiers

Chemical Name or Material A 1120
Assay Percent Range >99%
Molecular Formula C20H19F3N2O3
Formula Weight 392.37
Synonym 2-(4-(2-(Trifluoromethyl)phenyl)piperidine-1-carboxamido)benzoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

379310

 
r & d systems
RDS379310
10mg Each for $214.12

379350

 
r & d systems
RDS379350
50mg Each for $891.94
Description & Specifications

Specifications

Formula Weight 392.37
Quantity 10mg
Synonym 2-(4-(2-(Trifluoromethyl)phenyl)piperidine-1-carboxamido)benzoic acid
Recommended Storage Store at Room Temperature
Assay Percent Range >99%
Chemical Name or Material A 1120
CAS 1152782-19-8
Molecular Formula C20H19F3N2O3
Safety and Handling

Store at Room Temperature