Learn More
Thermo Scientific™ 9mm Autosampler Inserts
Clear glass or polypropylene inserts for 9mm HPLC and GC autosampler vials are available in a variety of configurations with usable volumes from 160μL to 3.5mL.
$77.15 - $1,783.43
Specifications
Diameter (Metric) Inner | 6 mm |
---|---|
Color | Clear |
Diameter (Metric) Outer | 6 mm |
Vial Type | Insert |
Certifications/Compliance | Certificate of Analysis (COA), Certificate of Conformance (COC) |
Catalog Number | Mfr. No. | Volume (Metric) | Material | Insert Type | Features | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (Metric) | Material | Insert Type | Features | Price | Quantity | ||
03-377-68
|
Thermo Scientific™
C4010627L |
400 μL | Glass | Pulled Point | Precision-formed tip, rests on vial bottom |
Pack of 100 Each for $123.70
Case of 10 Pack for $1,237.00 |
|
03-375-24C
|
Thermo Scientific™
C4010629 |
375 μL | Glass | Conical | Preferred for viscous solutions |
Pack of 100 Each for $170.60
Case of 10 Pack for $1,706.00 |
|
03-377-69
|
Thermo Scientific™
C4010629L |
400 μL | Glass | Precision Point | Precision-formed tip, rests on vial bottom |
Case of 1000 Each for $1,783.43
|
|
03-375-24AA
|
Thermo Scientific™
C4010629P |
325 μL | Polypropylene | Conical, Graduated | Conical Base; Graduation Marks |
Pack of 100 Each for $77.15
|
|
03-375-3A
![]() |
Thermo Scientific™
4025BS629DEL |
375 μL | Glass | Conical | Polyspring centers and cushions insert |
Case of 100 Each for $171.00
|
|
03-375-3B
![]() |
Thermo Scientific™
C4011631 |
500 μL | Glass | Flat Bottom | Insert rests on bottom of vial |
Pack of 500 Each for $186.74
|
![]() |
03-375-24CS
|
Thermo Scientific™
C4010S629 |
375 μL | Glass | Conical, Silanized | Insert rests on bottom of vial |
Pack of 100 Each for $230.55
|
|
03-375-3AS
|
Thermo Scientific™
C4010S630 |
375 μL | Glass | Conical, Silanized | Precision-formed tip, NSC silanized |
Pack of 100 Each for $372.21
|
|
03-375-3BS
|
Thermo Scientific™
C4011S631 |
500 μL | Glass | Flat Bottom, Silanized | Insert rests on bottom of vial |
Pack of 500 Each for $472.62
|
|
03-375-18E
|
Thermo Scientific™
C4010630P |
325 μL | Polypropylene | Conical | Precision-formed tip with Polyspring |
Pack of 100 Each for $125.23
Case of 10 Pack for $1,252.30 |
|
Description
Clear glass or polypropylene inserts for 9mm HPLC and GC autosampler vials are available in a variety of configurations with usable volumes from 160μL to 3.5mL.
- Pulled point inserts are an economical choice for noncritical applications
- Nominal O.D. of 6mm to fit Target wide-opening vials
For use with:
- Target LoVial Wide-Opening vials
- Snap-It™ Wide-Opening Snap Cap vials
- Target DP™ vials
- Target Wide-Opening vials
Specifications
6 mm | |
6 mm | |
Certificate of Analysis (COA), Certificate of Conformance (COC) |
Clear | |
Insert | |
Autosampler |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.