Please login to your online account to display your discounted pricing

9H-Purin-2-amine, 97%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC276560010

 View more versions of this product

Catalog No. AC276560010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 135.13
Physical Form Amorphous Powder
Quantity 1g
Color Light beige
Beilstein 26, 414
Melting Point 280° to 284°C
CAS Max % 100.0
Packaging Glass bottle
Chemical Name or Material 2-Aminopurine
CAS 452-06-2
Molecular Formula C5H5N5
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
CAS Min % 97.5
Synonym 1H-Purine-2-amine
Infrared Spectrum Authentic
Assay Percent Range 98%
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 2-Aminopurine
CAS 452-06-2
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C5H5N5
Formula Weight 135.13
Synonym 1H-Purine-2-amine
Accessories

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.

SDS & Safety Documents