missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Aminopurine, 98%, Thermo Scientific™

$264.94 - $264.94

Chemical Identifiers

CAS 452-06-2
Molecular Formula C5H5N5
Molecular Weight (g/mol) 135.13
MDL Number MFCD00005566
InChI Key MWBWWFOAEOYUST-UHFFFAOYSA-N
Synonym 1h-purin-2-amine, 2'-amino-purine, 2-amino purine, 2-aminopurine, 9h-purin-2-amine, 9h-purin-2-ylamine, aminopurine, ccris 759, purine, 2-amino, unii-o14b3u97fw
PubChem CID 9955
ChEBI CHEBI:479072
IUPAC Name 7H-purin-2-amine
SMILES NC1=NC=C2NC=NC2=N1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC276560500
SDS View Documents Promotion Details
Thermo Fisher Scientific
AC276560500
50mg Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC276562500
SDS View Documents Promotion Details
Thermo Fisher Scientific
276562500
250mg Glass bottle
Each for $264.94
 
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

452-06-2
135.13
MWBWWFOAEOYUST-UHFFFAOYSA-N
9955
7H-purin-2-amine
C5H5N5
MFCD00005566
1h-purin-2-amine, 2'-amino-purine, 2-amino purine, 2-aminopurine, 9h-purin-2-amine, 9h-purin-2-ylamine, aminopurine, ccris 759, purine, 2-amino, unii-o14b3u97fw
CHEBI:479072
NC1=NC=C2NC=NC2=N1

Specifications

452-06-2
100.0
280.0°C to 284.0°C
Authentic
MFCD00005566
Glass bottle
50mg
NC1=NC=C2NC=NC2=N1
135.13
CHEBI:479072
Amorphous Powder
2-Aminopurine
97.5
Beige
97.5% min
C5H5N5
26, 414
1h-purin-2-amine, 2'-amino-purine, 2-amino purine, 2-aminopurine, 9h-purin-2-amine, 9h-purin-2-ylamine, aminopurine, ccris 759, purine, 2-amino, unii-o14b3u97fw
MWBWWFOAEOYUST-UHFFFAOYSA-N
7H-purin-2-amine
9955
135.13
98%
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Product Certifications