missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Aminopurine, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 276562500

 View more versions of this product

Catalog No. AC276562500



Specifications

Specifications

2-Aminopurine
Glass bottle
280°C to 284°C
452-06-2
100.0
C5H5N5
1h-purin-2-amine, 2'-amino-purine, 2-amino purine, 2-aminopurine, 9h-purin-2-amine, 9h-purin-2-ylamine, aminopurine, ccris 759, purine, 2-amino, unii-o14b3u97fw
C1=C2C(=NC(=N1)N)N=CN2
135.13
CHEBI:479072
Amorphous Powder
Authentic
Beige
250mg
97.5
98%
26, 414
MWBWWFOAEOYUST-UHFFFAOYSA-N
7H-purin-2-amine
9955
135.13
≥97.5%

Chemical Identifiers

452-06-2
135.13
1h-purin-2-amine, 2'-amino-purine, 2-amino purine, 2-aminopurine, 9h-purin-2-amine, 9h-purin-2-ylamine, aminopurine, ccris 759, purine, 2-amino, unii-o14b3u97fw
CHEBI:479072
C1=C2C(=NC(=N1)N)N=CN2
C5H5N5
MWBWWFOAEOYUST-UHFFFAOYSA-N
9955
7H-purin-2-amine
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents