missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

9H-Fluorene-4-carboxylic acid, 96%, ACROS Organics™

Chemical Identifiers

CAS 6954-55-8
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 96%
Molecular Formula C14H10O2
Formula Weight 210.23
MDL Number MFCD00001112
Chemical Name or Material 9H-Fluorene-4-carboxylic acid
Assay ≥95%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC334020010 SDS Thermo Fisher Scientific
334020010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Chemical Identifiers

6954-55-8
100.0
C14H10O2
MFCD00001112
≥95%
95.0
96%
210.23
9H-Fluorene-4-carboxylic acid

Specifications

Powder
1g
190° to 195°C
100.0
96%
MFCD00001112
Glass bottle
9H-Fluorene-4-carboxylic acid
210.23
Beige
09, 690
95.0
C14H10O2
6954-55-8
Authentic
≥95%