Please login to your online account to display your discounted pricing

9H-Fluorene-4-carboxylic acid, 96%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  334020010

 View more versions of this product

Catalog No. AC334020010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥95%
Physical Form Powder
Formula Weight 210.23
Quantity 1g
Color Beige
Melting Point 190° to 195°C
Beilstein 09, 690
CAS Min % 95.0
CAS Max % 100.0
Chemical Name or Material 9H-Fluorene-4-carboxylic acid
Assay Percent Range 96%
Packaging Glass bottle
Molecular Formula C14H10O2
MDL Number MFCD00001112
CAS 6954-55-8
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 9H-Fluorene-4-carboxylic acid
CAS 6954-55-8
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C14H10O2
Formula Weight 210.23
MDL Number MFCD00001112