missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

9h-fluorene-1-carboxylic Acid, ACROS Organics™

Chemical Identifiers

Molecular Formula C14H10O2
Formula Weight 210.23
MDL Number MFCD00001130
Chemical Name or Material 9H-Fluorene-1-carboxylic acid
Assay ≥96.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC119180010 SDS Thermo Fisher Scientific
AC119180010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC119180100 SDS Thermo Fisher Scientific
AC119180100
Glass bottle 10g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

C14H10O2
MFCD00001130
≥96.0%
210.23
9H-Fluorene-1-carboxylic acid

Specifications

Light brown
1g
Authentic
Powder
210.23
09, 690
≥96.0%
246° to 250°C
6276-03-5
Glass bottle
C14H10O2
MFCD00001130
9H-Fluorene-1-carboxylic acid

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.